4-(Hydroxymethyl)pyridine-2-carboxylic acid 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-(Hydroxymethyl)picolinic acid
H32921.03
H32921.06
H32921.14
H32921.03EA
1480
CZK
H32921.03
H32921.06
H32921.14
4-(Hydroxymethyl)pyridine-2-carboxylic acid 95%
4-(Hydroxymethyl)pyridine-2-carboxylic acid
Purity: 95%
CAS No.: 923169-37-3
Molecular Formula: C7H7NO3
Formula Weight: 153.15
Melting Point: 178-185° dec.
MDL No.: MFCD08447083
Vzorec:
C₇H₇NO₃ MW: 153,15 g/mol Bod tání: 178…185 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD08447083 CAS číslo: 923169-37-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...