Tisk…

(6-Chloro-3-pyridinyl)methanol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 6-Chloropyridine-3-methanol, 2-Chloro-5-hydroxymethylpyridine, 6-Chloro-3-(hydroxymethyl)pyridine

M-Clarity™ quality level = MQ100

536016-25G 536016-5G
SIAL536016-25GEA 5170 CZK
SIAL536016-25G SIAL536016-5G
(6-Chloro-3-pyridinyl)methanol, Sigma-Aldrich®
(6-Chloro-3-pyridinyl)methanol
Vzorec: C₆H₆ClNO
MW: 143,57 g/mol
MDL Number: MFCD01075226
CAS číslo: 21543-49-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR