(6-Chloro-3-pyridinyl)methanol ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
L19285.06
L19285.14
L19285.06EA
2910
CZK
L19285.06
L19285.14
(6-Chloro-3-pyridinyl)methanol ≥98%
(6-Chloro-3-pyridinyl)methanol
Purity: 98%
CAS No.: 21543-49-7
Molecular Formula: C6H6ClNO
Formula Weight: 143.57
Melting Point: 46-49°
Boiling Point: 120°/0.4mm
MDL No.: MFCD01075226
Vzorec:
C₆H₆ClNO Bod varu: 120 °C (0,4 mmHg) Bod tání: 46…49 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075226 CAS číslo: 21543-49-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...