Tisk…

2-(4-Bromophenoxy)tetrahydropyran ≥98%

Dodavatel: Thermo Fisher Scientific

Synonyms: 2-(4-Bromophenoxy)tetrahydro-2H-pyran

H25914.09 H25914.18
H25914.09EA 1640 CZK
H25914.09 H25914.18
2-(4-Bromophenoxy)tetrahydropyran ≥98%
2-(4-Bromophenoxy)tetrahydropyran

Purity: 98% CAS No.: 36603-49-3 Molecular Formula: C11H13BrO2 Formula Weight: 257.13 Melting Point: 55-58° MDL No.: MFCD00091551

Vzorec: C₁₁H₁₃BrO₂
Bod tání: 55…58 °C
Storage Temperature: Ambient
MDL Number: MFCD00091551
CAS číslo: 36603-49-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR