Tisk…

4-Fluoro-1-naphthonitrile ≥97%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

B25335.14 B25335.06 B25335.03
B25335.14EA 11120 CZK
B25335.14 B25335.06 B25335.03
4-Fluoro-1-naphthonitrile ≥97%
4-Fluoro-1-naphthonitrile

Purity: 97% CAS No.: 13916-99-9 Molecular Formula: C11H6FN Formula Weight: 171.18 Melting Point: 89-91° UN No.: 3439 MDL No.: MFCD00236656

Vzorec: C₁₁H₆FN
Bod tání: 89…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00236656
CAS číslo: 13916-99-9
UN: 3439
ADR: 6.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR