Tisk…

1-Naphthonitrile 95%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

L14498.06 L14498.14
L14498.06EA 1100 CZK
L14498.06 L14498.14
1-Naphthonitrile 95%
1-Naphthonitrile

Purity: 95% CAS No.: 86-53-3 Molecular Formula: C11H7N Formula Weight: 153.18 Melting Point: 33-38° Boiling Point: 299° UN No.: 3439 MDL No.: MFCD00003866

Vzorec: C₁₁H₇N
Bod varu: 299 °C
Bod tání: 33…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00003866
CAS číslo: 86-53-3
EINECS: 201-679-8
UN: 3439
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR