Tisk…
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate), Sigma-Aldrich®
Dodavatel: Merck
Varování
16670-100MG 16670-1G 16670-5G-F
SIAL16670-100MGEA 4420 CZK
SIAL16670-100MG SIAL16670-1G SIAL16670-5G-F
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate), Sigma-Aldrich®
5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt (BCIP (p-toluidine salt), X-Phosphate)
Vzorec: C₁₅H₁₅BrClN₂O₄P
Storage Temperature: Freezer
MDL Number: MFCD00040636
CAS číslo: 6578-06-9
EINECS: 229-506-1