Tisk…

5-Bromo-4-chloro-3-indolyl phosphate dipotassium salt (BCIP-K), Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Phosphoric acid-mono-(5-bromo-4-chloro-1H-indol-3-yl) ester dipotassium salt

B6274-100MG B6274-1G B6274-25MG B6274-500MG B6274-50MG
SIALB6274-100MGEA 5940 CZK
SIALB6274-100MG SIALB6274-1G SIALB6274-25MG SIALB6274-500MG SIALB6274-50MG
5-Bromo-4-chloro-3-indolyl phosphate dipotassium salt (BCIP-K), Sigma-Aldrich®
5-Bromo-4-chloro-3-indolyl phosphate dipotassium salt (BCIP-K)
Vzorec: C₈H₄BrClK₂NO₄P
MW: 402,65 g/mol
MDL Number: MFCD00080893
CAS číslo: 102185-49-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR