Tricalcium diarsenate ≥95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Calcium arsenate
39219.18
39219.30
39219.18EA
4190
CZK
39219.18
39219.30
Tricalcium diarsenate ≥95%
Tricalcium diarsenate
Purity: 95%
CAS No.: 7778-44-1
Molecular Formula: Ca3(AsO4)2
Formula Weight: 398.03
Physical Form: -80 Mesh Powder
Density: 3.62
Appearance: White
Odour: Odourless
UN No.: 1573
MDL No.: MFCD00049407
Vzorec:
As₂Ca₃O₈ MW: 398,072 g/mol Hustota: 3,62 Storage Temperature: Ambient |
MDL Number:
MFCD00049407 CAS číslo: 7778-44-1 EINECS: 231-904-5 UN: 1573 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...