Tisk…

Potassium tetrakis(4-chlorophenyl)borate, Supelco®

Dodavatel: Merck

Synonyms: Potassiumtetrakis(4-chlorophenyl)borate

M-Clarity™ quality level = MQ100

60591-100MG 60591-1G 60591-5G
60591-100MGEA 1290 CZK
60591-100MG 60591-1G 60591-5G
Potassium tetrakis(4-chlorophenyl)borate, Supelco®
Potassium tetrakis(4-chlorophenyl)borate
Vzorec: C₂₄H₁₆BCl₄K
MW: 496,12 g/mol
MDL Number: MFCD00043105
CAS číslo: 14680-77-4
EINECS: 238-723-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45e6e569be36c0f5ab555bd4eb0a8607ae6fa4e9&vid=704f4ca5e17bb4ea957a11c42e4415351c2f59a7&prid=web&clid=SAPDAM

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR