Benzylphosphonic acid ≥97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
A13850.03
A13850.06
A13850.14
A13850.03EA
1040
CZK
A13850.03
A13850.06
A13850.14
Benzylphosphonic acid ≥97%
Benzylphosphonic acid
Purity: 97%
CAS No.: 6881-57-8
Molecular Formula: C6H5CH2PO3H2
Formula Weight: 172.11
Melting Point: 166-172°
Beilstein Reference: 2519318
UN No.: 3261
MDL No.: MFCD00039519
Vzorec:
C₇H₉O₃P MW: 172,11 g/mol Bod tání: 166…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039519 CAS číslo: 6881-57-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...