Tisk…

Diethyl benzylphosphonate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

D91071-100G D91071-25G
SIALD91071-100GEA 6150 CZK
SIALD91071-100G SIALD91071-25G
Diethyl benzylphosphonate, Sigma-Aldrich®
Diethyl benzylphosphonate
Vzorec: C₁₁H₁₇O₃P
MDL Number: MFCD00009078
CAS číslo: 1080-32-6
EINECS: 214-097-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR