Tisk…

Diethyl benzylphosphonate ≥99%

Dodavatel: Thermo Fisher Scientific
Varování

A10645.0B A10645.18 A10645.30
A10645.0BEA 40290 CZK
A10645.0B A10645.18 A10645.30
Diethyl benzylphosphonate ≥99%
Diethyl benzylphosphonate

Purity: 99% CAS No.: 1080-32-6 Molecular Formula: C11H17O3P Formula Weight: 228.23 Flash Point: 110°(230°F) Refractive Index: 1.4975 Boiling Point: 106-108°/1mm Beilstein Reference: 2580931 MDL No.: MFCD00009078

Vzorec: C₁₁H₁₇O₃P
Bod varu: 106…108 °C (1 mmHg)
Hustota: 1,094
Bod vzplanutí: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009078
CAS číslo: 1080-32-6
EINECS: 214-097-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR