Tisk…

5-Fluorobenzo[b]thiophene-2-carboxylic acid 96%

Dodavatel: Thermo Scientific
Varování

Synonyms: 5-Fluoro-1-benzothiophene-2-carboxylic acid

H33239.09 H33239.18 H33239.03 H33239.06
H33239.09EA 0 CZK
H33239.09 H33239.18 H33239.03 H33239.06
5-Fluorobenzo[b]thiophene-2-carboxylic acid 96%
5-Fluorobenzo[b]thiophene-2-carboxylic acid

Purity: 96% CAS No.: 70060-13-8 Molecular Formula: C9H5FO2S Formula Weight: 196.19 Melting Point: 257-262° MDL No.: MFCD01927184

Vzorec: C₉H₅FO₂S
MW: 196,19 g/mol
Bod tání: 257…262 °C
Storage Temperature: Ambient
MDL Number: MFCD01927184
CAS číslo: 70060-13-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR