3-Chlorobenzo[b]thiophene-2-carbonyl chlorid 95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
3-Chlorobenzo[b]thiophene-2-carbonyl chloride
L11745.03
L11745.06
L11745.03EA
1060
CZK
L11745.03
L11745.06
3-Chlorobenzo[b]thiophene-2-carbonyl chlorid 95%
3-Chlorobenzo[b]thiophene-2-carbonyl chlorid
Purity: 95%
CAS No.: 21815-91-8
Molecular Formula: C9H4Cl2OS
Formula Weight: 231.10
Melting Point: 111-114°
Beilstein Reference: 1309703
UN No.: 3261
MDL No.: MFCD00053069
Vzorec:
C₉H₄Cl₂OS MW: 231,10 g/mol Bod tání: 111…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053069 CAS číslo: 21815-91-8 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...