Terephthalic acid dihydrazide 95%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Terephthalic Dihydrazide
L01017.06
L01017.14
L01017.06EA
1020
CZK
L01017.06
L01017.14
Terephthalic acid dihydrazide 95%
Terephthalic acid dihydrazide
Purity: 95%
CAS No.: 136-64-1
Molecular Formula: C8H10N4O2
Formula Weight: 194.19
Melting Point:
Vzorec:
C₈H₁₀N₄O₂ MW: 194,19 g/mol Bod tání: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014762 CAS číslo: 136-64-1 EINECS: 205-253-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...