Tisk…

N,N-Diethylhydroxylamine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

471593-100ML 471593-2L 471593-500ML
SIAL471593-100MLEA 1290 CZK
SIAL471593-100ML SIAL471593-2L SIAL471593-500ML
N,N-Diethylhydroxylamine, Sigma-Aldrich®
N,N-Diethylhydroxylamine
Vzorec: C₄H₁₁NO
MW: 89,14 g/mol
MDL Number: MFCD00002126
CAS číslo: 3710-84-7
EINECS: 223-055-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR