Iminodiacetic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2,2'-Azanediyldiacetic acid
M-Clarity™ quality level = MQ200
56781-1KG
56781-250G
56781-50G
SIAL56781-1KGEA
5580
CZK
SIAL56781-1KG
SIAL56781-250G
SIAL56781-50G
Iminodiacetic acid, Sigma-Aldrich®
Iminodiacetic acid
Vzorec:
C₄H₇NO₄ MW: 133,10 g/mol |
MDL Number:
MFCD00004280 CAS číslo: 142-73-4 EINECS: 205-555-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...