Tisk…

Choline acetate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

670189-50G 670189-5G
SIAL670189-50GEA 17160 CZK
SIAL670189-50G SIAL670189-5G
Choline acetate, Sigma-Aldrich®
Choline acetate
Vzorec: C₆H₁₅NO₃
CAS číslo: 14586-35-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR