Tisk…

1-Acetyl-2-phenylhydrazine, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2-Phenylacetohydrazide, N-Acetyl-N'-phenylhydrazine

M-Clarity™ quality level = MQ100

A4626-25G A4626-100G
SIALA4626-25GEA 3020 CZK
SIALA4626-25G SIALA4626-100G
1-Acetyl-2-phenylhydrazine, Sigma-Aldrich®
1-Acetyl-2-phenylhydrazine
Vzorec: C₈H₁₀N₂O
MW: 150,18 g/mol
MDL Number: MFCD00008672
CAS číslo: 114-83-0
EINECS: 204-055-3
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR