Tisk…

1-Acetyl-2-phenylhydrazine ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

Synonyms: 2-Phenylacetohydrazide, N-Acetyl-N'-phenylhydrazine

A14446.14 A14446.22 A14446.36
A14446.14EA 1680 CZK
A14446.14 A14446.22 A14446.36
1-Acetyl-2-phenylhydrazine ≥98%
1-Acetyl-2-phenylhydrazine

Purity: 98% CAS No.: 114-83-0 Molecular Formula: C6H5NHNHCOCH3 Formula Weight: 150.18 Melting Point: 128-131° Beilstein Reference: 742880 UN No.: 2811 MDL No.: MFCD00008672

Vzorec: C₈H₁₀N₂O
MW: 150,18 g/mol
Bod tání: 128…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00008672
CAS číslo: 114-83-0
EINECS: 204-055-3
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR