Tisk…

Carbohydrazide ≥97%

Dodavatel: Thermo Scientific
Varování

A11145.14 A11145.22 A11145.36
A11145.14EA 1500 CZK
A11145.14 A11145.22 A11145.36
Carbohydrazide ≥97%
Carbohydrazide

Purity: 97% CAS No.: 497-18-7 Molecular Formula: CH6N4O Formula Weight: 90.09 Storage Temperature: 2°C to 8°C Melting Point: 153-158° UN No.: 3077 MDL No.: MFCD00007591

Vzorec: CH₆N₄O
MW: 90,09 g/mol
Bod tání: 153…158 °C
MDL Number: MFCD00007591
CAS číslo: 497-18-7
EINECS: 207-837-2
ADR: 9,III
Merck Index: 14,1804

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR