Tisk…

Acetoxyacetyl chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Acetoxyacetylchloride, 2-Chloro-2-oxoethyl acetate

M-Clarity™ quality level = MQ100

302368-25G 302368-5G
SIAL302368-25GEA 3300 CZK
SIAL302368-25G SIAL302368-5G
Acetoxyacetyl chloride, Sigma-Aldrich®
Acetoxyacetyl chloride
Vzorec: C₄H₅ClO₃
MDL Number: MFCD00011535
CAS číslo: 13831-31-7
EINECS: 237-542-4
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR