Tisk…

Aluminium acetate basic hydrate

Dodavatel: Thermo Fisher Scientific

Synonyms: Diacetoxy(hydroxy)aluminum

A11620.0E A11620.22 A11620.36 AB113376.0500
A11620.0EEA 14010 CZK
A11620.0E A11620.22 A11620.36 ABCRAB113376.0500
Aluminium acetate basic hydrate
Aluminium acetate basic hydrate

CAS No.: 142-03-0 Molecular Formula: approx (CH3CO2)2AlOH.xH2O Physical Form: Powder Appearance: White Merck Reference: 14,344 MDL No.: MFCD00008688

Vzorec: C₄H₇AlO₅·H₂O
MW: 162.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008688
CAS číslo: 142-03-0
EINECS: 205-518-2
Merck Index: 14,344

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR