Tisk…

Iminodiacetic acid disodium salt hydrate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

234877-100G 234877-500G
SIAL234877-100GEA 3830 CZK
SIAL234877-100G SIAL234877-500G
Iminodiacetic acid disodium salt hydrate, Sigma-Aldrich®
Iminodiacetic acid disodium salt hydrate
Vzorec: C₄H₅NNa₂O₄·xH₂O
MDL Number: MFCD00150689
CAS číslo: 207398-95-6
EINECS: 213-181-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR