Tisk…

Iminodiacetic acid disodium salt hydrate 97%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: Iminodiacetic acid disodium salt hydrate

43809.30 43809.18
43809.30EA 6700 CZK
43809.30 43809.18
Iminodiacetic acid disodium salt hydrate 97%
Iminodiacetic acid disodium salt hydrate

Purity: 97% CAS No.: 17593-73-6 Molecular Formula: HN(CH2CO2Na)2.xH2O Formula Weight: 177.07(anhy) Physical Form: Crystalline

Vzorec: C₄H₅NNa₂O₄·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00150689
CAS číslo: 17593-73-6
EINECS: 213-181-8
Merck Index: 14,4917

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR