Tisk…

Phthalan ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

A10217.06 A10217.14 A10217.22
A10217.06EA 1370 CZK
A10217.06 A10217.14 A10217.22
Phthalan ≥98%
Phthalan

Purity: 98% CAS No.: 496-14-0 Molecular Formula: C8H8O Formula Weight: 120.15 Flash Point: 63°(145°F) Refractive Index: 1.5460 Boiling Point: 192° Beilstein Reference: 111932 MDL No.: MFCD00005932

Vzorec: C₈H₈O
Bod varu: 192 °C
Hustota: 1,094
Bod vzplanutí: 63 °C (145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005932
CAS číslo: 496-14-0
EINECS: 207-815-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR