Tisk…

4-Methylumbelliferone, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-MU

M1381-100G M1381-25G
SIALM1381-100GEA 2240 CZK
SIALM1381-100G SIALM1381-25G
4-Methylumbelliferone, Sigma-Aldrich®
4-Methylumbelliferone
Vzorec: C₁₀H₈O₃
MW: 176,17 g/mol
MDL Number: MFCD00006866
CAS číslo: 90-33-5
EINECS: 201-986-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR