trans-4-Nitrocinnamaldehyde, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
trans-3-(4-Nitrophenyl)-2-propenal, (E)-3-(4-Nitrophenyl)acrylaldehyde, (E)-4-Nitrocinnamaldehyde, (E)-3-(4-Nitrophenyl)-2-propenal
M-Clarity™ quality level = MQ100
281670-1G
281670-5G
SIAL281670-1GEA
1190
CZK
SIAL281670-1G
SIAL281670-5G
trans-4-Nitrocinnamaldehyde, Sigma-Aldrich®
trans-4-Nitrocinnamaldehyde
Vzorec:
C₉H₇NO₃ MW: 177,16 g/mol |
MDL Number:
MFCD00007379 CAS číslo: 49678-08-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...