Tisk…

Pinacolyl methylphosphonate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

386588-1G 386588-5G
SIAL386588-1GEA 2660 CZK
SIAL386588-1G SIAL386588-5G
Pinacolyl methylphosphonate, Sigma-Aldrich®
Pinacolyl methylphosphonate
Vzorec: C₇H₁₇O₃P
CAS číslo: 616-52-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR