Tisk…

(Methyl)triphenoxyphosphonium iodide, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Methyltriphenoxyphosphonium iodide

M-Clarity™ quality level = MQ200

226432-10G 226432-50G
SIAL226432-10GEA 1440 CZK
SIAL226432-10G SIAL226432-50G
(Methyl)triphenoxyphosphonium iodide, Sigma-Aldrich®
(Methyl)triphenoxyphosphonium iodide
Vzorec: C₁₉H₁₈IO₃P
MDL Number: MFCD00011911
CAS číslo: 17579-99-6
EINECS: 241-551-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR