Tisk…

Phosphonoacetic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

A12117.09 A12117.14 A12117.22
A12117.09EA 3280 CZK
A12117.09 A12117.14 A12117.22
Phosphonoacetic acid ≥98%
Phosphonoacetic acid

Purity: 98+% CAS No.: 4408-78-0 Molecular Formula: C2H5O5P Formula Weight: 140.04 Odour: Odourless Melting Point: 139-146° Beilstein Reference: 1764355 UN No.: 3261 MDL No.: MFCD00004311

Vzorec: C₂H₅O₅P
MW: 140.04 g/mol
Bod tání: 139…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00004311
CAS číslo: 4408-78-0
EINECS: 224-558-1
UN: 3261
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR