Tisk…

Diethyl (cyanomethyl)phosphonate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

D91705-10G D91705-50G
SIALD91705-10GEA 2250 CZK
SIALD91705-10G SIALD91705-50G
Diethyl (cyanomethyl)phosphonate, Sigma-Aldrich®
Diethyl (cyanomethyl)phosphonate
Vzorec: C₆H₁₂NO₃P
MW: 177.14 g/mol
MDL Number: MFCD00001893
CAS číslo: 2537-48-6
EINECS: 219-806-0
UN: 3278
ADR: 6.1 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR