Tisk…

1-(Trimethylsiloxy)-1,3-butadiene, Sigma-Aldrich®

Dodavatel: Merck

227226-10G 227226-50G
SIAL227226-10GEA 5630 CZK
SIAL227226-10G SIAL227226-50G
1-(Trimethylsiloxy)-1,3-butadiene, Sigma-Aldrich®
1-(Trimethylsiloxy)-1,3-butadiene
Vzorec: C₇H₁₄OSi
CAS číslo: 6651-43-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR