5-Bromo-2'-deoxyuridine (broxuridine) ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Bromo-2'-deoxyuridine, 5-BrdU
H27260.03
H27260.06
H27260.MD
H27260.03EA
3780
CZK
H27260.03
H27260.06
H27260.MD
5-Bromo-2'-deoxyuridine (broxuridine) ≥99%
5-Bromo-2'-deoxyuridine (broxuridine)
Purity: 99%
CAS No.: 59-14-3
Molecular Formula: C9H11BrN2O5
Formula Weight: 307.10
Storage Temperature: 2°C to 8°C
Melting Point: ca 190° dec.
Beilstein Reference: 30395
Merck Reference: 14,1452
Optical Rotation: +31° (c=1 in 0.1N HCl)
MDL No.: MFCD00006529
Vzorec:
C₉H₁₁BrN₂O₅ |
MDL Number:
MFCD00006529 CAS číslo: 59-14-3 EINECS: 200-415-9 Merck Index: 14,1452 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...