Tisk…

Benzyloxyacetyl chloride, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

301019-25G 301019-5G
SIAL301019-25GEA 8000 CZK
SIAL301019-25G SIAL301019-5G
Benzyloxyacetyl chloride, Sigma-Aldrich®
Benzyloxyacetyl chloride
Vzorec: C₉H₉ClO₂
MDL Number: MFCD00010768
CAS číslo: 19810-31-2
UN: 3265
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR