Tisk…

N,N-Diisopropylcarbamoyl chloride 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

314520050 314520250
ACRO314520050EA 1800 CZK
ACRO314520050 ACRO314520250
N,N-Diisopropylcarbamoyl chloride 98%
N,N-Diisopropylcarbamoyl chloride
Vzorec: C₇H₁₄ClNO
Bod varu: 90…93 °C (15 mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD00015013
CAS číslo: 19009-39-3
EINECS: 242-743-5
UN: 3261
ADR: 8,II

Objednat

Výsledky specifikačního testu

Appearance Colorless to white to light yellow Crystalline powder or low melting solid
Infrared spectrum Conforms
Melting point 55 to 60 °C
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR