Tisk…

2,4-Dichloro-5-methoxypyrimidine, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

679089-10G 679089-1G
SIAL679089-10GEA 4670 CZK
SIAL679089-10G SIAL679089-1G
2,4-Dichloro-5-methoxypyrimidine, Sigma-Aldrich®
2,4-Dichloro-5-methoxypyrimidine
Vzorec: C₅H₄Cl₂N₂O
MW: 179,00 g/mol
MDL Number: MFCD02685670
CAS číslo: 19646-07-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR