(Aminomethyl)phosphonic acid, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
(Aminomethyl)phosphonic Acid, AMPA
M-Clarity™ quality level = MQ100
324817-1G
324817-250MG
SIAL324817-1GEA
9940
CZK
SIAL324817-1G
SIAL324817-250MG
(Aminomethyl)phosphonic acid, Sigma-Aldrich®
(Aminomethyl)phosphonic acid
Vzorec:
CH₆NO₃P MW: 111,04 g/mol |
MDL Number:
MFCD00008105 CAS číslo: 1066-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...