Tisk…

(Aminomethyl)phosphonic acid 99%

Dodavatel: Thermo Fisher Scientific
Varování

L09833.03 L09833.MD L09833.06
L09833.03EA 4880 CZK
L09833.03 L09833.MD L09833.06
(Aminomethyl)phosphonic acid 99%
(Aminomethyl)phosphonic acid

Purity: 99% CAS No.: 1066-51-9 Molecular Formula: CH6NO3P Formula Weight: 111.04 Melting Point: ca 290° dec. Beilstein Reference: 1701219

Vzorec: CH₆NO₃P
Storage Temperature: Ambient
MDL Number: MFCD00008105
CAS číslo: 1066-51-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR