Tisk…

(Aminomethyl)phosphonic acid 99%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: (Aminomethyl)phosphonic Acid, AMPA

291640010 291642500 291640050
ACRO291640010EA 5010 CZK
ACRO291640010 ACRO291642500 ACRO291640050
(Aminomethyl)phosphonic acid 99%
(Aminomethyl)phosphonic acid
Vzorec: CH₆NO₃P
Storage Temperature: Ambient
MDL Number: MFCD00008105
CAS číslo: 1066-51-9
UN: 3261
ADR: 8,III

Objednat

Výsledky specifikačního testu

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR