Perfluorobutane-1-sulphonyl fluoride 92%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Perfluoro-1-butanesulphonyl fluoride, Nonafluorobutanesulphonyl fluoride
312340250
312340050
312341000
ACRO312340250EA
3030
CZK
ACRO312340250
ACRO312340050
ACRO312341000
Perfluorobutane-1-sulphonyl fluoride 92%
Perfluorobutane-1-sulphonyl fluoride
Vzorec:
C₄F₁₀O₂S MW: 302,09 g/mol Bod varu: 64 °C Hustota: 1,68 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00007422 CAS číslo: 375-72-4 EINECS: 206-792-6 UN: 3265 ADR: 8,II |
Výsledky specifikačního testu
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
GC | ≥90 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...