Tisk…

8-Amino-2-naphthol 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 1-Amino-7-naphthol

A19404.03 A19404.06 A19404.14 A19404.A3 A19404.A1 A19404.A7
A19404.03EA 879 CZK
A19404.03 A19404.06 A19404.14 A19404.A3 A19404.A1 A19404.A7
8-Amino-2-naphthol 98%
8-Amino-2-naphthol

Purity: 98% CAS No.: 118-46-7 Molecular Formula: C10H9NO Formula Weight: 159.19 Melting Point: 206-210° MDL No.: MFCD00004031

Vzorec: C₁₀H₉NO
MW: 159,19 g/mol
Bod tání: 206…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00004031
CAS číslo: 118-46-7
EINECS: 204-252-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR