5-Amino-1-naphthol, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
376469-100G
376469-25G
SIAL376469-100GEA
3930
CZK
SIAL376469-100G
SIAL376469-25G
5-Amino-1-naphthol, Sigma-Aldrich®
5-Amino-1-naphthol
Vzorec:
C₁₀H₉NO |
MDL Number:
MFCD00041826 CAS číslo: 83-55-6 EINECS: 201-486-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...