Tisk…

Ethyl-4-chloroacetoacetate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Ethyl 4-chloro-3-oxobutanoate

M-Clarity™ quality level = MQ100

180769-250G 180769-50G
SIAL180769-250GEA 3080 CZK
SIAL180769-250G SIAL180769-50G
Ethyl-4-chloroacetoacetate, Sigma-Aldrich®
Ethyl-4-chloroacetoacetate
Vzorec: C₆H₉ClO₃
MW: 164,59 g/mol
MDL Number: MFCD00000939
CAS číslo: 638-07-3
EINECS: 211-317-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR