Thulium(III) chloride hexahydrate, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
204668-1G
204668-5G
SIAL204668-1GEA
6260
CZK
SIAL204668-1G
SIAL204668-5G
Thulium(III) chloride hexahydrate, Sigma-Aldrich®
Thulium(III) chloride hexahydrate
Vzorec:
Cl₃Tm·6H₂O |
CAS číslo:
1331-74-4 EINECS: 236-904-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...