Tisk…

Diphenylacetyl chloride ≥90%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2,2-Diphenylacetyl chloride

L00171.09 L00171.18
L00171.09EA 786 CZK
L00171.09 L00171.18
Diphenylacetyl chloride ≥90%
Diphenylacetyl chloride

Purity: 90+% CAS No.: 1871-76-7 Molecular Formula: C14H11ClO Formula Weight: 230.69 Melting Point: 50-56° Boiling Point: 175-176°/17mm Merck Reference: 14,3316 UN No.: 3261 MDL No.: MFCD00013655

Vzorec: C₁₄H₁₁ClO
Bod varu: 175…176 °C (17 mmHg)
Bod tání: 50…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00013655
CAS číslo: 1871-76-7
EINECS: 217-493-5
UN: 3261
ADR: 8,II
Merck Index: 14,3316

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR