Tisk…

1,3-Di-tert-butylimidazolium tetrafluoroborate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

659983-1G 659983-5G
SIAL659983-1GEA 1960 CZK
SIAL659983-1G SIAL659983-5G
1,3-Di-tert-butylimidazolium tetrafluoroborate, Sigma-Aldrich®
1,3-Di-tert-butylimidazolium tetrafluoroborate
Vzorec: C₁₁H₂₁BF₄N₂
MDL Number: MFCD07784425
CAS číslo: 263163-17-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR