Tisk…

6-Methoxy-2-naphthol, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 6-Methoxynaphthalen-2-ol, Naproxen Impurity H

65125-25G-F 65125-5G-F
SIAL65125-25G-FEA 16420 CZK
SIAL65125-25G-F SIAL65125-5G-F
6-Methoxy-2-naphthol, Sigma-Aldrich®
6-Methoxy-2-naphthol
Vzorec: C₁₁H₁₀O₂
MW: 174,2000 g/mol
MDL Number: MFCD00088677
CAS číslo: 5111-66-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR