Tetramethyl orthocarbonate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Tetramethylorthocarbonate
M-Clarity™ quality level = MQ200
132624-25G
132624-5G
SIAL132624-25GEA
26880
CZK
SIAL132624-25G
SIAL132624-5G
Tetramethyl orthocarbonate, Sigma-Aldrich®
Tetramethyl orthocarbonate
Vzorec:
C₅H₁₂O₄ MW: 136,15 g/mol |
MDL Number:
MFCD00008473 CAS číslo: 1850-14-2 EINECS: 217-438-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...